Anti-MYBPC1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT2944AEA 350 EUR
ABGEAT2944A
Anti-MYBPC1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MYBPC1 (myosin binding protein C, slow type)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human