Anti-NOVA1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3082AEA 350 EUR
ABGEAT3082A
Anti-NOVA1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: NOVA1 (neuro-oncological ventral antigen 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human