Anti-PALM2 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3177AEA 350 EUR
ABGEAT3177A
Anti-PALM2 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PALM2 (paralemmin 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human