Anti-PIGO Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3307AEA 350 EUR
ABGEAT3307A
Anti-PIGO Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PIGO (phosphatidylinositol glycan anchor biosynthesis, class O)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human