Anti-PIK3C2B Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAT3309AEA 350 EUR
ABGEAT3309A
Anti-PIK3C2B Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PIK3C2B (phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR