Anti-PIR Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3317AEA 350 EUR
ABGEAT3317A
Anti-PIR Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PIR (pirin (iron-binding nuclear protein))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human