Anti-PMS1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3357AEA 350 EUR
ABGEAT3357A
Anti-PMS1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PMS1 (PMS1 postmeiotic segregation increased 1 (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human