Anti-PTN Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3492AEA 350 EUR
ABGEAT3492A
Anti-PTN Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PTN (pleiotrophin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human