Anti-RARB Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3572AEA 350 EUR
ABGEAT3572A
Anti-RARB Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RARB (retinoic acid receptor, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human