Anti-RNASEH1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3652AEA 350 EUR
ABGEAT3652A
Anti-RNASEH1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RNASEH1 (ribonuclease H1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human