Anti-RP6-213H19.1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3697AEA 350 EUR
ABGEAT3697A
Anti-RP6-213H19.1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RP6-213H19.1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human