Anti-SERPINB2 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3821AEA 350 EUR
ABGEAT3821A
Anti-SERPINB2 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SERPINB2 (serpin peptidase inhibitor, clade B (ovalbumin), member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human