Anti-SH2D1A Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT3862AEA 350 EUR
ABGEAT3862A
Anti-SH2D1A Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SH2D1A (SH2 domain containing 1A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human