Anti-SSR4 Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAT4047AEA 350 EUR
ABGEAT4047A
Anti-SSR4 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SSR4 (signal sequence receptor, delta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR