Anti-STC1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4072AEA 350 EUR
ABGEAT4072A
Anti-STC1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: STC1 (stanniocalcin 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human