Anti-TCP1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4198AEA 350 EUR
ABGEAT4198A
Anti-TCP1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TCP1 (t-complex 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human