Anti-TWSG1 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4413AEA 350 EUR
ABGEAT4413A
Anti-TWSG1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TWSG1 (twisted gastrulation BMP signaling modulator 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human