Anti-UBE2Q2 Mouse Monoclonal Antibody

Toimittaja: Abgent
ABGEAT4442AEA 350 EUR
ABGEAT4442A
Anti-UBE2Q2 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: UBE2Q2 (ubiquitin-conjugating enzyme E2Q family member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR