Anti-UBE4A Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4446AEA 350 EUR
ABGEAT4446A
Anti-UBE4A Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: UBE4A (ubiquitination factor E4A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human