Anti-USP39 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4489AEA 350 EUR
ABGEAT4489A
Anti-USP39 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: USP39 (ubiquitin specific peptidase 39)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human