Anti-VAPA Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4501AEA 350 EUR
ABGEAT4501A
Anti-VAPA Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: VAPA (VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human