Anti-VIPR2 Mouse Monoclonal Antibody
Toimittaja: Abgent
ABGEAT4514AEA 350 EUR
ABGEAT4514A
Anti-VIPR2 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: VIPR2 (vasoactive intestinal peptide receptor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human