Anti-KLC2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP12241AEA 348 EUR
ABGEAP12241A
Anti-KLC2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: KLC2 (kinesin light chain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR