Anti-DDK Tag Monoclonal Antibody (PE (Phycoerythrin))

Toimittaja: OriGene
ORIGTA150020EA 476 EUR
ORIGTA150020
Anti-DDK Tag Monoclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Type: Tag
Antigen: DDK Tag
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR