Anti-HA tag None Polyclonal Antibody

Toimittaja: OriGene
ORIGTA150034EA 620 EUR
ORIGTA150034
Anti-HA tag None Polyclonal Antibody
Vasta-aineet
Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR