Anti-mPlum Mouse Monoclonal Antibody

Toimittaja: OriGene
ORIGTA180072EA 497 EUR
ORIGTA180072
Anti-mPlum Mouse Monoclonal Antibody
Vasta-aineet
Type: Tag
Antigen: mPlum
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR