Anti-PKM Mouse Monoclonal Antibody [clone: 4E8]

Toimittaja: OriGene
ORIGTA190162EA 465 EUR
ORIGTA190162
Anti-PKM Mouse Monoclonal Antibody [clone: 4E8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: Monoclonal
Clone:4E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR