Anti-LEO1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA300199EA 697 EUR
ORIGTA300199
Anti-LEO1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: LEO1 (Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR