Anti-KRT5 Rabbit Monoclonal Antibody [clone: EP1601Y]

Toimittaja: OriGene
ORIGTA300887EA 714 EUR
ORIGTA300887
Anti-KRT5 Rabbit Monoclonal Antibody [clone: EP1601Y]
Recommended Dilutions: Western Blot: 1:10000

Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR