Anti-CCND1 Rabbit monoclonal antibody [clone: EPR2241]

Toimittaja: OriGene
ORIGTA301123EA 714 EUR
ORIGTA301123
Anti-CCND1 Rabbit monoclonal antibody [clone: EPR2241]
Recommended Dilutions: Western Blot: 1:10000; Immunoprecipitation: 1:40; Immunofluorescence: 1:100; Immunochemistry: 1:100

Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR