Anti-ACTN1 Rabbit Monoclonal Antibody [clone: EP2527Y]

Toimittaja: OriGene
ORIGTA301137EA 716 EUR
ORIGTA301137
Anti-ACTN1 Rabbit Monoclonal Antibody [clone: EP2527Y]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1: 100; Flow Cytometry: 1:80; Immunoprecipitation: 1:40

Type: Primary
Antigen: ACTN1 (actinin, alpha 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR