Anti-NGF Rabbit Monoclonal Antibody [clone: EP1318Y]

Toimittaja: OriGene
ORIGTA301201EA 666 EUR
ORIGTA301201
Anti-NGF Rabbit Monoclonal Antibody [clone: EP1318Y]
Vasta-aineet
Recommended Dilutions: Western Blot: 1: 1000; Immunochemistry: 1: 100; Flow Cytometry: 1:200; Immunoprecipitation: 1:20

Type: Primary
Antigen: NGF (nerve growth factor (beta polypeptide))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR