Anti-EPS15L1 Rabbit Monoclonal Antibody [clone: EP1146Y]

Toimittaja: OriGene
ORIGTA301237EA 566 EUR
ORIGTA301237
Anti-EPS15L1 Rabbit Monoclonal Antibody [clone: EP1146Y]
Recommended Dilutions: Western Blot: 1:10000-1:20000; Flow Cytometry: 1:20; Immunochemistry/Immunofluorescence: 1:100-1:250

Type: Primary
Antigen: EPS15L1 (epidermal growth factor receptor pathway substrate 15-like 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR