Anti-PTGDS Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301421EA 601 EUR
ORIGTA301421
Anti-PTGDS Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot:2 ?g/ml

Type: Primary
Antigen: PTGDS (prostaglandin D2 synthase 21kDa (brain))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR