Anti-EPAS1 Mouse Monoclonal Antibody [clone: ep190b]

Toimittaja: OriGene
ORIGTA301435EA 683 EUR
ORIGTA301435
Anti-EPAS1 Mouse Monoclonal Antibody [clone: ep190b]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1: 150-1:300

Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR