Anti-ARNT Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301442EA 747 EUR
ORIGTA301442
Anti-ARNT Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ARNT (aryl hydrocarbon receptor nuclear translocator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR