Anti-UCHL1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301477EA 624 EUR
ORIGTA301477
Anti-UCHL1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR