Anti-SIRT6 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301524EA 569 EUR
ORIGTA301524
Anti-SIRT6 Rabbit Polyclonal Antibody
Vasta-aineet
  • Recommended Dilutions: Western Blot: 2μg/ml

Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR