Anti-MAP1LC3B Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301543EA 747 EUR
ORIGTA301543
Anti-MAP1LC3B Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:3000

Type: Primary
Antigen: MAP1LC3B (microtubule-associated protein 1 light chain 3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR