Anti-LPIN1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301606EA 569 EUR
ORIGTA301606
Anti-LPIN1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: LPIN1 (lipin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR