Anti-ATRIP Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301615EA 624 EUR
ORIGTA301615
Anti-ATRIP Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:5000-1: 15000

Type: Primary
Antigen: ATRIP (ATR interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR