Anti-PSMD10 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301722EA 569 EUR
ORIGTA301722
Anti-PSMD10 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: PSMD10 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR