Anti-AHR Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301733EA 658 EUR
ORIGTA301733
Anti-AHR Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: AHR (aryl hydrocarbon receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR