Anti-SPECC1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301749EA 569 EUR
ORIGTA301749
Anti-SPECC1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: SPECC1 (sperm antigen with calponin homology and coiled-coil domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR