Anti-APEX1 Mouse Monoclonal Antibody [clone: 13B8E5C2]

Toimittaja: OriGene
ORIGTA301765EA 747 EUR
ORIGTA301765
Anti-APEX1 Mouse Monoclonal Antibody [clone: 13B8E5C2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:5000

Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR