Anti-ARNTL Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301766EA 624 EUR
ORIGTA301766
Anti-ARNTL Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1: 100-1:200

Type: Primary
Antigen: ARNTL (aryl hydrocarbon receptor nuclear translocator-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR