Anti-RAD9A Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301769EA 484 EUR
ORIGTA301769
Anti-RAD9A Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1: 1000-1:5000; Immunochemistry: 1:800

Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR