Anti-PLTP Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA301774EA 516 EUR
ORIGTA301774
Anti-PLTP Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PLTP (phospholipid transfer protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR