Anti-SLC29A1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302009EA 637 EUR
ORIGTA302009
Anti-SLC29A1 Rabbit Polyclonal Antibody
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SLC29A1 (solute carrier family 29 (equilibrative nucleoside transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR