Anti-SOX2 Mouse Monoclonal Antibody

Toimittaja: OriGene
ORIGTA302025EA 698 EUR
ORIGTA302025
Anti-SOX2 Mouse Monoclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 1:200-2000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR